siRNA Details
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278041.1 | NP_001264970.1 | 6 | 724 | ||
| NM_009893.2 | NP_034023.1 | 6 | 658 | |||
| GenBank mRNA | AF069501.1 | - | 6 | 658 | ||
| AF096276.1 | - | 6 | 637 | |||
| AK077460.1 | - | 6 | 654 | |||
| AK149445.1 | - | 6 | 633 | |||
| AK165339.1 | - | 6 | 651 | |||
| BC145017.1 | - | 6 | 722 | |||
| BC145018.1 | - | 6 | 722 | |||
| BC145019.1 | - | 6 | 656 | |||
| BC145696.1 | - | 6 | 722 | 40130999 | ||

Chrd
Genomic Map