siRNA Details
| Gene Symbol: Clgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009904.2 | NP_034034.2 | 10 | 1253 | ||
| GenBank mRNA | AK019534.1 | - | 10 | 1167 | ||
| AK034525.1 | - | 10 | 1154 | |||
| BC050767.1 | - | 10 | 1253 | 6771060 | ||
| D14117.1 | - | 10 | 1266 | |||
| D86323.1 | - | 10 | 1266 | |||
| U08373.1 | - | 10 | 1266 | |||

Clgn
Genomic Map