siRNA Details
| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB003174.1 | - | 2 | 696 | ||
| AF045146.1 | - | 1 | 607 | |||
| BC096626.1 | - | 2 | 669 | 30749408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcr3| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB003174.1 | - | 2 | 696 | ||
| AF045146.1 | - | 1 | 607 | |||
| BC096626.1 | - | 2 | 669 | 30749408 | ||