siRNA Details
| Gene Symbol: HAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005328.2 | NP_005319.1 | 2, 3 | 1173 | ||
| GenBank mRNA | AK297724.1 | - | 2, 3 | 1219 | ||
| BC069353.1 | - | 2, 3 | 707 | 7262337 | ||
| BC109071.1 | - | 2, 3 | 1078 | 40034202 | ||
| BC109072.1 | - | 2, 3 | 1078 | 40034203 | ||
| U54804.1 | - | 2, 3 | 1170 | |||

HAS2
Genomic Map