siRNA Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009922.4 | NP_034052.3 | 5 | 593 | ||
| GenBank mRNA | AK035548.1 | - | 5 | 566 | ||
| BC138863.1 | - | 5 | 525 | 8861885 | ||
| BC138864.1 | - | 5 | 525 | 8861886 | ||
| BC145446.1 | - | 5 | 525 | |||
| Z19542.1 | - | 5 | 587 | |||

Cnn1
Genomic Map