siRNA Details
| Gene Symbol: Cnr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036658.1 | - | 2 | 1802 | ||
| AK037898.1 | - | 2 | 1912 | |||
| AK134109.1 | - | 2 | 1838 | |||
| BC024052.1 | - | 2 | 1767 | 5098695 | ||
| X86405.1 | - | 2 | 1755 | |||
| X93168.1 | - | 2 | 1807 | |||

Cnr2
Genomic Map