siRNA Details
| Gene Symbol: Copa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009938.4 | NP_034068.3 | 13 | 1540 | ||
| GenBank mRNA | AK043479.1 | - | 13 | 1266 | ||
| AK200292.1 | - | 2 | 83 | |||
| BC024070.1 | - | 13 | 1193 | 5097228 | ||
| BC047429.1 | - | 13 | 1231 | 3498656 | ||
| BC082785.1 | - | 9 | 880 | |||

Copa
Genomic Map