siRNA Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF053770.1 | - | 1 | 137 | ||
| AF080580.1 | - | 2 | 200 | |||
| AF098949.1 | - | 2 | 266 | |||
| AK137841.1 | - | 2 | 264 | |||
| AK158380.1 | - | 2 | 251 | |||
| BC038681.1 | - | 2 | 233 | 1380996 | ||
| BY284216.1 | - | 2 | 305 | |||

Coq7
Genomic Map