siRNA Details
| Gene Symbol: Cpne6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136057.2 | NP_001129529.1 | 5 | 735 | ||
| NM_001146183.1 | NP_001139655.1 | 5 | 712 | |||
| NM_009947.3 | NP_034077.1 | 5 | 538 | |||
| GenBank mRNA | AB008893.1 | - | 5 | 538 | ||
| AK134548.1 | - | 5 | 557 | |||
| AK144410.1 | - | 5 | 564 | |||
| AK158638.1 | - | 4 | 1957 | |||
| BC050766.1 | - | 5 | 560 | 6591063 | ||
| BC051994.1 | - | 4 | 1120 | |||

Cpne6
Genomic Map