siRNA Details
| Gene Symbol: Cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010012.3 | NP_034142.3 | 1 | 597 | ||
| GenBank mRNA | AF090317.1 | - | 1 | 583 | ||
| BC010973.1 | - | 1 | 607 | 4161038 | ||
| BC010974.1 | - | 1 | 600 | 4162964 | ||
| BC049969.1 | - | 1 | 582 | 4158774 | ||

Cyp8b1
Genomic Map