siRNA Details
| Gene Symbol: Dcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110222.1 | NP_001103692.1 | 8 | 2543 | ||
| NM_001110223.1 | NP_001103693.1 | 8 | 2465 | |||
| NM_001110224.1 | NP_001103694.1 | 7 | 2462 | |||
| NM_010025.2 | NP_034155.2 | 7 | 2447 | |||
| GenBank mRNA | AB011678.1 | - | 8 | 2563 | ||
| AK164119.1 | - | 7 | 2349 | |||
| AY560329.1 | - | 8 | 2354 | |||
| BC056391.1 | - | 7 | 2356 | 6855006 | ||
| BC057010.1 | - | 7 | 2448 | 6825750 | ||
| BC062974.1 | - | 7 | 2448 | 6825750 | ||

Dcx
Genomic Map