siRNA Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010049.3 | NP_034179.1 | 6 | 1309 | ||
| GenBank mRNA | AK031241.1 | - | 6 | 943 | ||
| AK136212.1 | - | 6 | 1367 | |||
| BC005796.1 | - | 6 | 881 | 3707582 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhfr| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010049.3 | NP_034179.1 | 6 | 1309 | ||
| GenBank mRNA | AK031241.1 | - | 6 | 943 | ||
| AK136212.1 | - | 6 | 1367 | |||
| BC005796.1 | - | 6 | 881 | 3707582 | ||