siRNA Details
| Gene Symbol: Dio2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010050.2 | NP_034180.1 | 2 | 1318 | ||
| GenBank mRNA | AF093137.1 | - | 2 | 1176 | ||
| AF096875.1 | - | 2 | 1317 | |||
| AF177196.1 | - | 2 | 1188 | |||
| AK032038.1 | - | 2 | 1028 | |||
| AK045607.1 | - | 2 | 1072 | |||
| BC125383.1 | - | 2 | 1213 | 40129898 | ||
| BC125385.1 | - | 2 | 1213 | 40129900 | ||
| CV555526.1 | - | 1 | 351 | |||

Dio2
Genomic Map