siRNA Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301737.1 | NP_001288666.1 | 4 | 573 | ||
| NM_010065.3 | NP_034195.2 | 4 | 573 | |||
| GenBank mRNA | AK011651.1 | - | 4 | 540 | ||
| AK144142.1 | - | 4 | 524 | |||
| AK220483.1 | - | 4 | 515 | |||
| BC034679.1 | - | 4 | 527 | 4506865 | ||
| BC058623.1 | - | 4 | 507 | 5706803 | ||
| CA328097.1 | - | 2 | 199 | |||
| L31395.1 | - | 4 | 470 | |||
| L31397.1 | - | 4 | 526 | |||

Dnm1
Genomic Map