siRNA Details
| Gene Symbol: Dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010073.2 | NP_034203.1 | 4 | 377 | ||
| GenBank mRNA | AK002468.1 | - | 4 | 361 | ||
| AK004241.1 | - | 4 | 352 | |||
| BC008256.1 | - | 4 | 377 | 2648752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpm2| Gene Symbol: Dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010073.2 | NP_034203.1 | 4 | 377 | ||
| GenBank mRNA | AK002468.1 | - | 4 | 361 | ||
| AK004241.1 | - | 4 | 352 | |||
| BC008256.1 | - | 4 | 377 | 2648752 | ||