siRNA Details
| Gene Symbol: Adam24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010086.4 | NP_034216.3 | 2 | 1495 | ||
| GenBank mRNA | AF167402.1 | - | 1 | 1483 | ||
| AK029528.1 | - | 2 | 1496 | |||
| AK133199.1 | - | 2 | 1511 | |||
| BC145785.1 | - | 1 | 1384 | 40131088 | ||
| KF144358.1 | - | 1 | 115 | |||
| KF144359.1 | - | 1 | 115 | |||
| U06151.1 | - | 1 | 61 | |||

Adam24
Genomic Map