siRNA Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276387.1 | NP_001263316.1 | 3 | 1147 | ||
| NM_010095.6 | NP_034225.1 | 3 | 1142 | |||
| GenBank mRNA | AK053013.1 | - | 3 | 1143 | ||
| AK139262.1 | - | 3 | 854 | |||
| BC049188.1 | - | 2 | 441 | 6312260 | ||
| BC050922.1 | - | 2 | 664 | 6409737 | ||
| U71189.1 | - | 2 | 327 | |||
| U82441.1 | - | 2 | 196 | |||
| U92703.1 | - | 2 | 547 | |||

Ebf2
Genomic Map