siRNA Details
| Gene Symbol: Clcn4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035804.1 | - | 3 | 227 | ||
| AK133759.1 | - | 3 | 253 | |||
| AK191145.1 | - | 2 | 40 | |||
| AK209368.1 | - | 2 | 54 | |||
| BC005553.1 | - | 3 | 288 | 3586154 | ||
| BC110668.1 | - | 4 | 587 | 6391053 | ||
| BY035169.1 | - | 3 | 335 | |||
| DQ186662.1 | - | 2 | 85 | |||
| Z49916.1 | - | 4 | 498 | |||

Clcn4-2
Genomic Map