siRNA Details
| Gene Symbol: SPRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097589.1 | NP_001091058.1 | 2 | 188 | ||
| NM_005416.2 | NP_005407.1 | 3 | 272 | |||
| GenBank mRNA | AJ243667.1 | - | 2 | 170 | ||
| AK311823.1 | - | 2 | 186 | |||
| AY118269.1 | - | 2 | 164 | |||
| BC017802.1 | - | 2 | 186 | 4288753 | ||
| BF826427.1 | - | 3 | 274 | |||
| DA451130.1 | - | 3 | 272 | |||
| DA763011.1 | - | 2 | 186 | |||
| EF553525.1 | - | 2 | 1236 | |||

SPRR3
Genomic Map