siRNA Details
| Gene Symbol: Bop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013481.1 | NP_038509.1 | 6 | 743 | ||
| GenBank mRNA | AK079627.1 | - | 6 | 723 | ||
| AK144095.1 | - | 6 | 754 | |||
| AK167110.1 | - | 6 | 726 | |||
| AK167225.1 | - | 6 | 727 | |||
| AK167829.1 | - | 6 | 721 | |||
| AK167878.1 | - | 6 | 731 | |||
| AK168127.1 | - | 6 | 724 | |||
| BC012693.1 | - | 6 | 717 | 4193123 | ||
| U77415.1 | - | 6 | 743 | |||

Bop1
Genomic Map