siRNA Details
| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013486.2 | NP_038514.1 | 5 | 807 | ||
| GenBank mRNA | AF065906.1 | - | 5 | 804 | ||
| AF065907.1 | - | 5 | 804 | |||
| AF065908.1 | - | 5 | 804 | |||
| AF065909.1 | - | 5 | 804 | |||
| AF306543.1 | - | 5 | 804 | |||
| AK138728.1 | - | 4 | 558 | |||
| BC037185.1 | - | 2 | 642 | |||
| BC053731.1 | - | 5 | 790 | 30077134 | ||
| M18934.1 | - | 5 | 794 | |||
| X06143.1 | - | 5 | 762 | |||
| Y00023.1 | - | 5 | 807 | |||

Cd2
Genomic Map