siRNA Details
| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013487.3 | NP_038515.3 | 2 | 306 | ||
| GenBank mRNA | AK042172.1 | - | 2 | 308 | ||
| AK050629.1 | - | 2 | 295 | |||
| AK153843.1 | - | 2 | 271 | |||
| AK154064.1 | - | 2 | 268 | |||
| BC062807.1 | - | 2 | 211 | 1366111 | ||
| EF382667.1 | - | 2 | 206 | |||
| HM536173.1 | - | 2 | 206 | |||
| X02339.1 | - | 2 | 174 | |||

Cd3d
Genomic Map