siRNA Details
| Gene Symbol: Crem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110850.2 | NP_001104320.1 | 6 | 951 | ||
| NM_001110854.1 | NP_001104324.2 | 4 | 593 | |||
| NM_001110855.1 | NP_001104325.2 | 3 | 557 | |||
| NM_001110856.2 | NP_001104326.1 | 8 | 1134 | |||
| NM_001271504.1 | NP_001258433.1 | 7 | 1098 | |||
| NM_001271505.1 | NP_001258434.1 | 8 | 1176 | |||
| NM_001271506.1 | NP_001258435.1 | 8 | 1287 | |||
| NM_013498.3 | NP_038526.2 | 7 | 987 | |||
| GenBank mRNA | AY738716.1 | - | 5 | 867 | ||
| AY738717.1 | - | 6 | 1014 | |||
| AY738718.1 | - | 7 | 1050 | |||
| AY738719.1 | - | 7 | 1092 | |||
| AY738720.1 | - | 7 | 1110 | |||
| AY738721.1 | - | 7 | 1203 | |||
| M60285.1 | - | 7 | 915 | |||

Crem
Genomic Map