siRNA Details
| Gene Symbol: Ate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029895.3 | NP_001025066.1 | 3, 4 | 628 | ||
| NM_001136054.2 | NP_001129526.1 | 3, 4 | 360 | |||
| NM_001271343.1 | NP_001258272.1 | 3, 4 | 628 | |||
| NM_013799.3 | NP_038827.2 | 3, 4 | 360 | |||
| GenBank mRNA | AK138277.1 | - | 3, 4 | 279 | ||
| AK148442.1 | - | 3, 4 | 328 | |||
| AK158613.1 | - | 3, 4 | 630 | |||
| AK167591.1 | - | 3, 4 | 320 | |||
| BC042601.1 | - | 3, 4 | 319 | 4503326 | ||
| BC050948.1 | - | 3, 4 | 310 | 6826691 | ||
| DQ093639.1 | - | 3, 4 | 220 | |||
| DQ093640.1 | - | 3, 4 | 220 | |||

Ate1
Genomic Map