siRNA Details
| Gene Symbol: Atp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289445.1 | NP_001276374.1 | 28 | 3304 | ||
| NM_001289446.1 | NP_001276375.1 | 28 | 3034 | |||
| NM_015731.3 | NP_056546.2 | 28 | 3104 | |||
| GenBank mRNA | AF152243.1 | - | 28 | 3272 | ||
| AK082271.1 | - | 28 | 3030 | |||
| AK089064.1 | - | 28 | 3210 | |||
| BC006949.1 | - | 26 | 2806 | |||
| BC075718.1 | - | 28 | 3232 | |||
| BC094551.1 | - | 28 | 3074 | 6813369 | ||

Atp9a
Genomic Map