siRNA Details
| Gene Symbol: Atp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289445.1 | NP_001276374.1 | 28 | 3686 | ||
| NM_001289446.1 | NP_001276375.1 | 28 | 3416 | |||
| NM_015731.3 | NP_056546.2 | 28 | 3486 | |||
| GenBank mRNA | AK082271.1 | - | 28 | 3412 | ||
| AK089064.1 | - | 28 | 3591 | |||
| AK182263.1 | - | 1 | 47 | |||
| BC006949.1 | - | 26 | 3187 | |||
| BC075718.1 | - | 28 | 3613 | |||
| BC094551.1 | - | 28 | 3456 | 6813369 | ||

Atp9a
Genomic Map