siRNA Details
| Gene Symbol: CRYBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057093.1 | NP_476434.1 | 4 | 860 | ||
| NM_057094.1 | NP_476435.1 | 5 | 666 | |||
| GenBank mRNA | AF166331.1 | - | 5 | 660 | ||
| BC006285.2 | - | 4 | 860 | 3960073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYBA2| Gene Symbol: CRYBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057093.1 | NP_476434.1 | 4 | 860 | ||
| NM_057094.1 | NP_476435.1 | 5 | 666 | |||
| GenBank mRNA | AF166331.1 | - | 5 | 660 | ||
| BC006285.2 | - | 4 | 860 | 3960073 | ||