siRNA Details
| Gene Symbol: CSTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005213.3 | NP_005204.1 | 3 | 395 | ||
| GenBank mRNA | AK291308.1 | - | 3 | 311 | ||
| AV717631.1 | - | 3 | 305 | |||
| BC010379.1 | - | 3 | 315 | 4063525 | ||
| BG213814.1 | - | 2 | 202 | |||
| CR456914.1 | - | 3 | 266 | |||
| X05978.1 | - | 3 | 311 | |||

CSTA
Genomic Map