siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016704.2 | NP_057913.2 | 13 | 2000 | ||
| GenBank mRNA | AF184900.1 | - | 13 | 2001 | ||
| BC011251.1 | - | 13 | 1989 | 4196330 | ||
| CB948485.1 | - | 3 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016704.2 | NP_057913.2 | 13 | 2000 | ||
| GenBank mRNA | AF184900.1 | - | 13 | 2001 | ||
| BC011251.1 | - | 13 | 1989 | 4196330 | ||
| CB948485.1 | - | 3 | 377 | |||