siRNA Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016709.2 | NP_057918.2 | 8, 9 | 863 | ||
| GenBank mRNA | AK003219.1 | - | 8, 9 | 843 | ||
| BC026525.1 | - | 8, 9 | 801 | |||
| BC049597.1 | - | 8, 9 | 847 | 6593786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Auh| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016709.2 | NP_057918.2 | 8, 9 | 863 | ||
| GenBank mRNA | AK003219.1 | - | 8, 9 | 843 | ||
| BC026525.1 | - | 8, 9 | 801 | |||
| BC049597.1 | - | 8, 9 | 847 | 6593786 | ||