siRNA Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016811.2 | NP_058091.2 | 9 | 959 | ||
| GenBank mRNA | AF085219.1 | - | 8 | 645 | ||
| BC006713.1 | - | 9 | 959 | 3584592 | ||
| JN832677.1 | - | 10 | 991 | |||
| JN832678.1 | - | 9 | 978 | |||
| JN832679.1 | - | 9 | 978 | |||

Dgka
Genomic Map