siRNA Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016811.2 | NP_058091.2 | 18 | 1796 | ||
| GenBank mRNA | AF085219.1 | - | 17 | 1482 | ||
| AK008063.1 | - | 6 | 460 | |||
| AK008314.1 | - | 7 | 467 | |||
| BC006713.1 | - | 18 | 1796 | 3584592 | ||
| JN832677.1 | - | 18 | 1739 | |||
| JN832678.1 | - | 18 | 1844 | |||
| JN832679.1 | - | 18 | 1815 | |||

Dgka
Genomic Map