siRNA Details
| Gene Symbol: Gdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019506.4 | NP_062379.3 | 2 | 1051 | ||
| GenBank mRNA | AF156890.1 | - | 2 | 1051 | ||
| AK147029.1 | - | 2 | 1052 | |||
| BC103625.1 | - | 2 | 1020 | 40047137 | ||
| BC103679.1 | - | 2 | 1020 | 40047139 | ||
| BC103680.1 | - | 2 | 1020 | 40047138 | ||
| BC103681.1 | - | 2 | 1020 | 40047136 | ||

Gdf2
Genomic Map