siRNA Details
| Gene Symbol: Cbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019626.3 | NP_062600.2 | 3 | 1976 | ||
| GenBank mRNA | AK158818.1 | - | 3 | 1977 | ||
| AK158885.1 | - | 3 | 1994 | |||
| BC051956.1 | - | 3 | 1592 | |||
| BC055730.1 | - | 3 | 1680 | 6404555 | ||
| X61448.1 | - | 2 | 1494 | |||

Cbln1
Genomic Map