siRNA Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020260.2 | NP_064656.2 | 10 | 1445 | ||
| GenBank mRNA | AF151363.1 | - | 10 | 1086 | ||
| AK138417.1 | - | 10 | 1685 | |||
| AK149975.1 | - | 10 | 1281 | |||
| AK150109.1 | - | 10 | 1281 | |||
| AK155729.1 | - | 10 | 1265 | |||
| AK170685.1 | - | 10 | 1265 | |||
| BC138870.1 | - | 10 | 1250 | 8861892 | ||

Arhgap31
Genomic Map