siRNA Details
| Gene Symbol: Cryba4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021351.1 | NP_067326.1 | 2 | 159 | ||
| GenBank mRNA | AJ272228.2 | - | 2 | 159 | ||
| AK132591.1 | - | 3 | 176 | |||
| AK161601.1 | - | 1 | 123 | |||
| AK162018.1 | - | 3 | 173 | |||
| BC056444.1 | - | 2 | 149 | 6824277 | ||
| BC058703.1 | - | 2 | 159 | 6824293 | ||

Cryba4
Genomic Map