siRNA Details
| Gene Symbol: Drg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021354.1 | NP_067329.1 | 3 | 336 | ||
| GenBank mRNA | AJ243590.1 | - | 3 | 336 | ||
| BC082564.1 | - | 3 | 292 | 30551418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drg2| Gene Symbol: Drg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021354.1 | NP_067329.1 | 3 | 336 | ||
| GenBank mRNA | AJ243590.1 | - | 3 | 336 | ||
| BC082564.1 | - | 3 | 292 | 30551418 | ||