siRNA Details
| Gene Symbol: Drg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021354.1 | NP_067329.1 | 12 | 1054 | ||
| GenBank mRNA | AJ243590.1 | - | 12 | 1054 | ||
| AK012988.1 | - | 12 | 1022 | |||
| AK197050.1 | - | 1 | 41 | |||
| BC082564.1 | - | 12 | 1010 | 30551418 | ||

Drg2
Genomic Map