siRNA Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301230.1 | NP_001288159.1 | 7 | 2013 | ||
| NM_001301232.1 | NP_001288161.1 | 6 | 1906 | |||
| NM_023635.6 | NP_076124.1 | 6 | 1864 | |||
| GenBank mRNA | AF304376.1 | - | 6 | 1820 | ||
| AK008798.1 | - | 1 | 37 | |||
| AK010373.1 | - | 6 | 1782 | |||
| AK030529.1 | - | 6 | 1811 | |||
| AK207170.1 | - | 1 | 169 | |||
| AK208837.1 | - | 1 | 171 | |||
| AK219379.1 | - | 1 | 168 | |||

Rab27a
Genomic Map