siRNA Details
| Gene Symbol: Clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023671.2 | NP_076160.1 | 5 | 644 | ||
| GenBank mRNA | AK011789.1 | - | 5 | 631 | ||
| AK031877.1 | - | 5 | 626 | |||
| AK133981.1 | - | 5 | 637 | |||
| AK151356.1 | - | 5 | 629 | |||
| AK151944.1 | - | 5 | 629 | |||
| AK169259.1 | - | 5 | 608 | |||
| BC005575.1 | - | 5 | 610 | 3967933 | ||
| CX236388.1 | - | 5 | 644 | |||

Clns1a
Genomic Map