siRNA Details
| Gene Symbol: Tmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028953.2 | NP_083229.1 | 8 | 776 | ||
| GenBank mRNA | AF417579.1 | - | 8 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmc1| Gene Symbol: Tmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028953.2 | NP_083229.1 | 8 | 776 | ||
| GenBank mRNA | AF417579.1 | - | 8 | 776 | ||