siRNA Details
| Gene Symbol: Dsg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030596.3 | NP_085099.2 | 8 | 931 | ||
| GenBank mRNA | AK029018.1 | - | 953 | |||
| AK037142.1 | - | 8 | 962 | |||
| AK148657.1 | - | 8 | 932 | |||
| BC132058.1 | - | 8 | 960 | 40130335 | ||
| BC132639.1 | - | 8 | 960 | 40130916 | ||
| U86016.1 | - | 8 | 860 | |||

Dsg3
Genomic Map