siRNA Details
Gene Symbol: Chrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033269.4 | NP_150372.1 | 5 | 2692 | ||
GenBank mRNA | AK080950.1 | - | 5 | 2694 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
