siRNA Details
| Gene Symbol: Hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146323.1 | NP_001139795.1 | 4, 5 | 617 | ||
| NM_001146324.1 | NP_001139796.1 | 4, 5 | 1013 | |||
| NM_080634.4 | NP_542365.3 | 4, 5 | 1013 | |||
| GenBank mRNA | AF393747.1 | - | 4, 5 | 1002 | ||
| AF393780.1 | - | 4, 5 | 1005 | |||
| AK034377.1 | - | 4, 5 | 984 | |||
| AK036663.1 | - | 4, 5 | 1021 | |||
| AK037829.1 | - | 4, 5 | 512 | |||
| AK049204.1 | - | 4, 5 | 1006 | |||
| AK079896.1 | - | 4, 5 | 1005 | |||
| AK149359.1 | - | 4, 5 | 999 | |||
| AK155497.1 | - | 4, 5 | 980 | |||
| AK169855.1 | - | 3, 4 | 1085 | |||
| AK170581.1 | - | 4, 5 | 529 | |||
| AK171997.1 | - | 4, 5 | 605 | |||
| BC058235.1 | - | 4, 5 | 987 | 6307400 | ||

Hps3
Genomic Map