siRNA Details
| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111059.1 | NP_001104529.1 | 8 | 1678 | ||
| NM_133654.3 | NP_598415.1 | 8 | 1522 | |||
| GenBank mRNA | AK044121.1 | - | 8 | 1614 | ||
| AK053874.1 | - | 8 | 1458 | |||
| AK089010.1 | - | 8 | 1428 | |||
| AK167329.1 | - | 8 | 1589 | |||
| AK167350.1 | - | 8 | 1589 | |||
| BC006607.1 | - | 8 | 1438 | 3966337 | ||
| BC066820.1 | - | 8 | 1551 | |||

Cd34
Genomic Map