siRNA Details
| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277219.1 | NP_001264148.1 | 3 | 1883 | ||
| NM_001277221.1 | NP_001264150.1 | 2 | 1517 | |||
| NM_133656.5 | NP_598417.2 | 3 | 2053 | |||
| GenBank mRNA | AK028179.1 | - | 3 | 1861 | ||
| AK028488.1 | - | 3 | 2003 | |||
| AK036905.1 | - | 3 | 1834 | |||
| AK052795.1 | - | 3 | 1842 | |||
| AK076097.1 | - | 3 | 1748 | |||
| AK088519.1 | - | 3 | 1854 | |||
| AK139089.1 | - | 3 | 1839 | |||
| AK152549.1 | - | 3 | 2027 | |||
| AK154987.1 | - | 3 | 2036 | |||
| AK161761.1 | - | 3 | 1995 | |||
| BC012216.1 | - | 3 | 1989 | 3992341 | ||
| BC031149.1 | - | 3 | 1816 | 4975654 | ||

Crk
Genomic Map