siRNA Details
| Gene Symbol: Cobl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282993.1 | NP_001269922.1 | 2 | 471 | ||
| NM_001282994.1 | NP_001269923.1 | 2 | 302 | |||
| NM_172496.3 | NP_766084.3 | 2 | 471 | |||
| GenBank mRNA | AK028833.1 | - | 2 | 318 | ||
| AK049730.1 | - | 2 | 264 | |||
| AK122334.1 | - | 2 | 302 | |||
| AK147366.1 | - | 2 | 472 | |||
| AK147633.1 | - | 2 | 472 | |||
| AY308745.1 | - | 2 | 311 | |||
| BC145116.1 | - | 2 | 311 | |||
| BC145118.1 | - | 2 | 311 | 9053666 | ||
| BC150706.1 | - | 2 | 311 | 9087617 | ||
| U26967.1 | - | 1 | 44 | |||

Cobl
Genomic Map