siRNA Details
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175087.4 | NP_780296.1 | 5 | 1825 | ||
GenBank mRNA | AK082699.1 | - | 5 | 1830 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175087.4 | NP_780296.1 | 5 | 1825 | ||
GenBank mRNA | AK082699.1 | - | 5 | 1830 |