siRNA Details
| Gene Symbol: Camk2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039138.2 | NP_001034227.1 | 10 | 864 | ||
| NM_001039139.2 | NP_001034228.1 | 10 | 864 | |||
| NM_178597.5 | NP_848712.2 | 10 | 864 | |||
| GenBank mRNA | AF395884.1 | - | 10 | 741 | ||
| AK031213.1 | - | 10 | 827 | |||
| AK054406.1 | - | 9 | 736 | |||
| AK078311.1 | - | 10 | 808 | |||
| AK081969.1 | - | 9 | 1266 | |||
| AK154764.1 | - | 10 | 793 | |||
| BC019162.1 | - | 10 | 816 | 3675224 | ||
| BC025597.1 | - | 10 | 818 | 5342641 | ||

Camk2g
Genomic Map