siRNA Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198408.3 | NP_940800.1 | 7 | 1305 | ||
| GenBank mRNA | AK135067.1 | - | 7 | 1306 | ||
| AK139404.1 | - | 7 | 1151 | |||
| AK164173.1 | - | 1 | 315 | |||
| BC061247.1 | - | 7 | 1246 | 30246702 | ||

Crhbp
Genomic Map